Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLCG1
Entrez Gene ID
5335
Gene Name
phospholipase C gamma 1
Gene Aliases
NCKAP3, PLC-II, PLC1, PLC148, PLCgamma1
Chromosome Location
Chr.20: 41137519 - 41175721 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002660.2 NP_002651.2 7 - 8 836 69
NM_182811.1 NP_877963.1 7 - 8 836 69
XM_005260438.2 XP_005260495.1 7 - 8 838 69
XM_011528865.1 XP_011527167.1 7 - 8 703 69
XM_011528867.2 XP_011527169.1 7 - 8 836 69
GenBank mRNA AB210028.1 - 7 - 8 1120 69
BC136466.1 - 7 - 8 803 69
BC144136.1 - 7 - 8 803 69
M34667.1 - 7 - 8 791 69

View Full Product Details