The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204504.2 | NP_001191433.1 | 20 - 21 | 2500 | 128 |
NM_001204505.2 | NP_001191434.1 | 17 - 18 | 2121 | 128 | |
NM_001204506.2 | NP_001191435.1 | 17 - 18 | 2121 | 128 | |
NM_001297429.1 | NP_001284358.1 | 15 - 16 | 1888 | 128 | |
NM_152910.5 | NP_690874.2 | 19 - 20 | 2463 | 128 | |
NM_178009.4 | NP_821077.1 | 19 - 20 | 2463 | 128 | |
NR_123714.1 | 17 - 18 | 2121 | 128 | ||
NR_123715.1 | 20 - 21 | 2929 | 128 | ||
GenBank mRNA | AB078967.1 | - | 19 - 20 | 2418 | 128 |
AB078968.1 | - | 19 - 20 | 2418 | 128 | |
AK098302.1 | - | 4 - 5 | 482 | 128 | |
AK122909.1 | - | 17 - 18 | 2086 | 128 | |
AK289546.1 | - | 20 - 21 | 2502 | 128 | |
AK302631.1 | - | 15 - 16 | 1848 | 128 | |
AL833883.1 | - | 5 - 6 | 551 | 128 | |
BC043292.2 | - | 17 - 18 | 2078 | 128 |