The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204504.2 | NP_001191433.1 | 27 - 28 | 3313 | 81 |
NM_001204505.2 | NP_001191434.1 | 24 - 25 | 2934 | 81 | |
NM_001204506.2 | NP_001191435.1 | 24 - 25 | 2934 | 81 | |
NM_001297429.1 | NP_001284358.1 | 22 - 23 | 2701 | 81 | |
NM_152910.5 | NP_690874.2 | 26 - 27 | 3276 | 81 | |
NM_178009.4 | NP_821077.1 | 26 - 27 | 3276 | 81 | |
NR_123714.1 | 24 - 25 | 2934 | 81 | ||
NR_123715.1 | 27 - 28 | 3742 | 81 | ||
GenBank mRNA | AB078967.1 | - | 26 - 27 | 3231 | 81 |
AB078968.1 | - | 26 - 27 | 3231 | 81 | |
AK098302.1 | - | 11 - 12 | 1295 | 81 | |
AK122909.1 | - | 24 - 25 | 2899 | 81 | |
AK289546.1 | - | 27 - 28 | 3315 | 81 | |
AK302631.1 | - | 22 - 23 | 2661 | 81 | |
AK302727.1 | - | 24 - 25 | 2934 | 81 | |
AL833883.1 | - | 12 - 13 | 1364 | 81 | |
BC043292.2 | - | 24 - 25 | 2891 | 81 |