The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204504.2 | NP_001191433.1 | 5 - 6 | 596 | 91 |
NM_001204505.2 | NP_001191434.1 | 2 - 3 | 217 | 91 | |
NM_001204506.2 | NP_001191435.1 | 2 - 3 | 217 | 91 | |
NM_001297429.1 | NP_001284358.1 | 2 - 3 | 217 | 91 | |
NM_152910.5 | NP_690874.2 | 4 - 5 | 559 | 91 | |
NM_178009.4 | NP_821077.1 | 4 - 5 | 559 | 91 | |
NR_123714.1 | 2 - 3 | 217 | 91 | ||
NR_123715.1 | 4 - 5 | 830 | 91 | ||
GenBank mRNA | AB078967.1 | - | 4 - 5 | 514 | 91 |
AB078968.1 | - | 4 - 5 | 514 | 91 | |
AK122909.1 | - | 2 - 3 | 183 | 91 | |
AK289546.1 | - | 5 - 6 | 598 | 91 | |
AK302631.1 | - | 2 - 3 | 177 | 91 | |
AK302727.1 | - | 2 - 3 | 217 | 91 | |
BC043292.2 | - | 2 - 3 | 174 | 91 |