The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204504.2 | NP_001191433.1 | 7 - 8 | 836 | 88 |
NM_001204505.2 | NP_001191434.1 | 4 - 5 | 457 | 88 | |
NM_001204506.2 | NP_001191435.1 | 4 - 5 | 457 | 88 | |
NM_152910.5 | NP_690874.2 | 6 - 7 | 799 | 88 | |
NM_178009.4 | NP_821077.1 | 6 - 7 | 799 | 88 | |
NR_123714.1 | 4 - 5 | 457 | 88 | ||
NR_123715.1 | 6 - 7 | 1070 | 88 | ||
GenBank mRNA | AB078967.1 | - | 6 - 7 | 754 | 88 |
AB078968.1 | - | 6 - 7 | 754 | 88 | |
AK122909.1 | - | 4 - 5 | 423 | 88 | |
AK289546.1 | - | 7 - 8 | 838 | 88 | |
AK302727.1 | - | 4 - 5 | 457 | 88 | |
BC043292.2 | - | 4 - 5 | 414 | 88 |