The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204504.2 | NP_001191433.1 | 15 - 16 | 1735 | 73 |
NM_001204505.2 | NP_001191434.1 | 12 - 13 | 1356 | 73 | |
NM_001204506.2 | NP_001191435.1 | 12 - 13 | 1356 | 73 | |
NM_001297429.1 | NP_001284358.1 | 10 - 11 | 1123 | 73 | |
NM_152910.5 | NP_690874.2 | 14 - 15 | 1698 | 73 | |
NM_178009.4 | NP_821077.1 | 14 - 15 | 1698 | 73 | |
NR_123714.1 | 12 - 13 | 1356 | 73 | ||
NR_123715.1 | 14 - 15 | 1969 | 73 | ||
GenBank mRNA | AB078967.1 | - | 14 - 15 | 1653 | 73 |
AB078968.1 | - | 14 - 15 | 1653 | 73 | |
AK122909.1 | - | 12 - 13 | 1321 | 73 | |
AK289546.1 | - | 15 - 16 | 1737 | 73 | |
AK302631.1 | - | 10 - 11 | 1083 | 73 | |
AK302727.1 | - | 12 - 13 | 1356 | 73 | |
BC043292.2 | - | 12 - 13 | 1313 | 73 |