Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR11
Entrez Gene ID
55717
Gene Name
WD repeat domain 11
Gene Aliases
BRWD2, DR11, HH14, SRI1, WDR15
Chromosome Location
Chr.10: 120851175 - 120909526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK124522.1 - 6 - 7 2949 101

View Full Product Details