The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083907.2 | NP_001077376.2 | 6 - 7 | 1059 | 144 |
NM_001127507.2 | NP_001120979.2 | 5 - 6 | 891 | 144 | |
NM_017935.4 | NP_060405.4 | 6 - 7 | 1290 | 144 | |
XM_017008337.1 | XP_016863826.1 | 6 - 7 | 1037 | 144 | |
GenBank mRNA | AB063170.1 | - | 5 - 6 | 926 | 144 |
AK000713.1 | - | 6 - 7 | 1006 | 144 | |
AK091523.1 | - | 10 - 11 | 1391 | 144 | |
AL834291.1 | - | 6 - 7 | 1109 | 144 | |
BC032241.1 | - | 6 - 7 | 1158 | 144 | |
EU051376.1 | - | 5 - 6 | 629 | 144 |