The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152592.3 | NP_689805.3 | 12 - 13 | 2259 | 59 |
XM_005267377.3 | XP_005267434.1 | 13 - 14 | 2356 | 59 | |
XM_006720063.3 | XP_006720126.1 | 13 - 14 | 2456 | 59 | |
XM_011536515.2 | XP_011534817.1 | 13 - 14 | 2391 | 59 | |
XM_017021056.1 | XP_016876545.1 | 12 - 13 | 2258 | 59 | |
GenBank mRNA | AK098471.1 | - | 7 - 8 | 1022 | 59 |
AK131436.1 | - | 13 - 14 | 2124 | 59 | |
BC146604.1 | - | 13 - 14 | 2259 | 59 | |
BX648302.1 | - | 10 - 11 | 4131 | 59 |