The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178034.3 | NP_828848.3 | 9 - 10 | 874 | 126 |
XM_011521467.2 | XP_011519769.1 | 8 - 9 | 705 | 126 | |
GenBank mRNA | AB090876.1 | - | 9 - 10 | 877 | 126 |