Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location
Chr.12: 98729904 - 99984727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204065.1 NP_001190994.1 2 - 3 375 84
NM_001204068.1 NP_001190997.1 4 - 5 871 84
NM_001204079.1 NP_001191008.1 2 - 3 375 84
NM_001204080.1 NP_001191009.1 2 - 3 375 84
NM_001204081.1 NP_001191010.1 2 - 3 375 84
NM_152788.4 NP_690001.3 18 - 19 3303 84
NM_181670.3 NP_858056.2 4 - 5 871 84
XM_005269028.4 XP_005269085.1 18 - 19 2914 84
XM_005269029.4 XP_005269086.1 18 - 19 2874 84
XM_005269032.3 XP_005269089.1 2 - 3 239 84
XM_006719504.3 XP_006719567.1 18 - 19 3376 84
XM_006719505.3 XP_006719568.1 18 - 19 3376 84
XM_006719506.3 XP_006719569.1 18 - 19 3376 84
XM_006719507.3 XP_006719570.1 18 - 19 3376 84
XM_006719508.3 XP_006719571.1 18 - 19 3376 84
XM_006719509.3 XP_006719572.1 18 - 19 3376 84
XM_006719510.3 XP_006719573.1 18 - 19 3376 84
XM_017019651.1 XP_016875140.1 18 - 19 3376 84
XM_017019652.1 XP_016875141.1 18 - 19 3364 84
XM_017019653.1 XP_016875142.1 18 - 19 3376 84
XM_017019654.1 XP_016875143.1 18 - 19 3364 84
XM_017019655.1 XP_016875144.1 18 - 19 3376 84
XM_017019659.1 XP_016875148.1 4 - 5 867 84
XM_017019660.1 XP_016875149.1 4 - 5 867 84
XM_017019661.1 XP_016875150.1 5 - 6 581 84
XM_017019662.1 XP_016875151.1 5 - 6 711 84
XM_017019664.1 XP_016875153.1 5 - 6 523 84
GenBank mRNA AF145204.1 - 12 - 13 2007 84
AK289768.1 - 4 - 5 836 84
AK294994.1 - 2 - 3 247 84
AK295988.1 - 2 - 3 348 84
AK315954.1 - 2 - 3 230 84
AM980945.1 - 2 - 3 231 84
AY281131.1 - 18 - 19 2889 84
AY281132.1 - 4 - 5 564 84
AY356353.1 - 2 - 3 204 84
AY620824.1 - 9 - 10 1614 84
BC026313.1 - 4 - 5 766 84
BP193713.1 - 2 - 3 347 84

View Full Product Details