The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033503.2 | NP_001028675.1 | 7 - 8 | 726 | 77 |
NM_016103.3 | NP_057187.1 | 6 - 7 | 607 | 77 | |
XM_017009521.1 | XP_016865010.1 | 6 - 7 | 614 | 77 | |
GenBank mRNA | AF087850.1 | - | 6 - 7 | 644 | 77 |
AF092130.1 | - | 6 - 7 | 623 | 77 | |
AK056821.1 | - | 3 - 4 | 2441 | 77 | |
AK223452.1 | - | 6 - 7 | 592 | 77 | |
AL512710.1 | - | 6 - 7 | 1642 | 77 | |
BC002847.2 | - | 7 - 8 | 657 | 77 | |
BC093034.1 | - | 6 - 7 | 607 | 77 |