Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PARL
Entrez Gene ID
55486
Gene Name
presenilin associated rhomboid like
Gene Aliases
PRO2207, PSARL, PSARL1, PSENIP2, RHBDS1
Chromosome Location
Chr.3: 183826488 - 183884908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324436.1 NP_001311365.1 5 - 6 666 151
NM_001324438.1 NP_001311367.1 5 - 6 666 151
NM_018622.6 NP_061092.3 5 - 6 666 151
NR_136893.1 5 - 6 722 151
XM_005247582.4 XP_005247639.1 5 - 6 661 151
XM_017006800.1 XP_016862289.1 5 - 6 661 151
GenBank mRNA AF197937.1 - 5 - 6 604 151
AK024765.1 - 5 - 6 638 151
BC014058.2 - 5 - 6 624 151

View Full Product Details