The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146702.1 | NP_001140174.1 | 15 - 16 | 2845 | 118 |
NM_001282622.1 | NP_001269551.1 | 17 - 18 | 3043 | 118 | |
NM_004187.3 | NP_004178.2 | 17 - 18 | 3046 | 118 | |
XM_005262035.4 | XP_005262092.1 | 17 - 18 | 2859 | 118 | |
XM_011530824.2 | XP_011529126.1 | 17 - 18 | 2859 | 118 | |
XM_011530825.2 | XP_011529127.1 | 16 - 17 | 2736 | 118 | |
XM_011530826.2 | XP_011529128.1 | 16 - 17 | 2736 | 118 | |
XM_011530827.2 | XP_011529129.1 | 17 - 18 | 2859 | 118 | |
XM_011530828.1 | XP_011529130.1 | 17 - 18 | 2859 | 118 | |
XM_011530829.2 | XP_011529131.1 | 18 - 19 | 2928 | 118 | |
XM_011530830.2 | XP_011529132.1 | 18 - 19 | 2875 | 118 | |
XM_011530831.2 | XP_011529133.1 | 11 - 12 | 1867 | 118 | |
XM_017029886.1 | XP_016885375.1 | 17 - 18 | 2856 | 118 | |
XM_017029887.1 | XP_016885376.1 | 17 - 18 | 2859 | 118 | |
XM_017029888.1 | XP_016885377.1 | 17 - 18 | 2856 | 118 | |
XM_017029889.1 | XP_016885378.1 | 17 - 18 | 2859 | 118 | |
GenBank mRNA | AK304732.1 | - | 15 - 16 | 2628 | 118 |
AK308077.1 | - | 17 - 18 | 2823 | 118 | |
BC054499.2 | - | 17 - 18 | 2797 | 118 | |
EF613277.1 | - | 17 - 18 | 2513 | 118 | |
L25270.1 | - | 17 - 18 | 3044 | 118 |