The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282622.1 | NP_001269551.1 | 2 - 3 | 756 | 96 |
NM_004187.3 | NP_004178.2 | 2 - 3 | 756 | 96 | |
XM_005262035.4 | XP_005262092.1 | 2 - 3 | 569 | 96 | |
XM_011530824.2 | XP_011529126.1 | 2 - 3 | 569 | 96 | |
XM_011530827.2 | XP_011529129.1 | 2 - 3 | 569 | 96 | |
XM_011530828.1 | XP_011529130.1 | 2 - 3 | 569 | 96 | |
XM_011530829.2 | XP_011529131.1 | 3 - 4 | 638 | 96 | |
XM_011530830.2 | XP_011529132.1 | 3 - 4 | 585 | 96 | |
XM_017029886.1 | XP_016885375.1 | 2 - 3 | 569 | 96 | |
XM_017029887.1 | XP_016885376.1 | 2 - 3 | 569 | 96 | |
XM_017029888.1 | XP_016885377.1 | 2 - 3 | 569 | 96 | |
XM_017029889.1 | XP_016885378.1 | 2 - 3 | 569 | 96 | |
GenBank mRNA | AK308077.1 | - | 2 - 3 | 533 | 96 |
BC054499.2 | - | 2 - 3 | 510 | 96 | |
EF613277.1 | - | 2 - 3 | 223 | 96 | |
L25270.1 | - | 2 - 3 | 754 | 96 |