Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL4A4
Entrez Gene ID
1286
Gene Name
collagen type IV alpha 4 chain
Gene Aliases
CA44
Chromosome Location
Chr.2: 226970293 - 227164559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000092.4 NP_000083.3 26 - 27 2707 151
XM_005246281.3 XP_005246338.1 26 - 27 2241 151
XM_005246282.3 XP_005246339.1 20 - 21 1824 151
XM_006712246.3 XP_006712309.1 26 - 27 2379 151
XM_011510557.2 XP_011508859.1 26 - 27 2379 151
XM_011510559.2 XP_011508861.1 26 - 27 2379 151
XM_011510560.2 XP_011508862.1 26 - 27 2379 151
XM_011510561.2 XP_011508863.1 26 - 27 2379 151
XM_011510562.2 XP_011508864.1 26 - 27 2379 151
XM_011510565.2 XP_011508867.1 26 - 27 2379 151
XM_011510566.2 XP_011508868.1 26 - 27 2379 151
XM_011510567.2 XP_011508869.1 26 - 27 2379 151
XM_011510568.2 XP_011508870.1 26 - 27 2379 151
XM_011510569.2 XP_011508871.1 26 - 27 2379 151
XM_011510570.2 XP_011508872.1 26 - 27 2379 151
XM_011510572.2 XP_011508874.1 5 - 6 604 151
XM_017003296.1 XP_016858785.1 26 - 27 2379 151
XM_017003297.1 XP_016858786.1 26 - 27 2379 151
XM_017003298.1 XP_016858787.1 26 - 27 2379 151
XM_017003299.1 XP_016858788.1 26 - 27 2379 151
XM_017003300.1 XP_016858789.1 26 - 27 2379 151

View Full Product Details