Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL4A4
Entrez Gene ID
1286
Gene Name
collagen type IV alpha 4 chain
Gene Aliases
CA44
Chromosome Location
Chr.2: 226970293 - 227164559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000092.4 NP_000083.3 47 - 48 5470 83
XM_005246281.3 XP_005246338.1 47 - 48 5004 83
XM_005246282.3 XP_005246339.1 41 - 42 4587 83
XM_006712246.3 XP_006712309.1 46 - 47 4953 83
XM_011510557.2 XP_011508859.1 48 - 49 5061 83
XM_011510558.2 XP_011508860.1 46 - 47 5034 83
XM_011510572.2 XP_011508874.1 26 - 27 3367 83
XM_017003297.1 XP_016858786.1 47 - 48 5025 83
GenBank mRNA D17391.1 - 9 - 10 1163 83
X81053.1 - 47 - 48 5024 83

View Full Product Details