The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000466.2 | NP_000457.1 | 3 - 4 | 456 | 114 |
NM_001282677.1 | NP_001269606.1 | 3 - 4 | 456 | 114 | |
GenBank mRNA | AB008112.1 | - | 3 - 4 | 427 | 114 |
AB052090.1 | - | 3 - 4 | 360 | 114 | |
AB052091.1 | - | 3 - 4 | 360 | 114 | |
AB052092.1 | - | 3 - 4 | 360 | 114 | |
AB052093.1 | - | 3 - 4 | 360 | 114 | |
AB052094.1 | - | 3 - 4 | 360 | 114 | |
AF026086.1 | - | 3 - 4 | 420 | 114 | |
AF030356.1 | - | 3 - 4 | 364 | 114 | |
AK292955.1 | - | 3 - 4 | 361 | 114 | |
BC035575.1 | - | 3 - 4 | 398 | 114 | |
DA313282.1 | - | 3 - 4 | 457 | 114 |