Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DENND3
Entrez Gene ID
22898
Gene Name
DENN domain containing 3
Gene Aliases
-
Chromosome Location
Chr.8: 141128618 - 141195804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014957.2 NP_055772.2 17 - 18 2986 88
XM_005250838.4 XP_005250895.2 17 - 18 2877 88
XM_005250839.2 XP_005250896.2 17 - 18 2877 88
XM_005250840.4 XP_005250897.2 16 - 17 2721 88
XM_005250841.3 XP_005250898.2 16 - 17 2721 88
XM_005250842.4 XP_005250899.1 15 - 16 2738 88
XM_011516933.1 XP_011515235.1 17 - 18 2877 88
XM_011516934.2 XP_011515236.1 17 - 18 2877 88
XM_011516937.1 XP_011515239.1 17 - 18 2877 88
XM_011516938.2 XP_011515240.1 10 - 11 2032 88
XM_011516939.2 XP_011515241.1 8 - 9 1555 88
XM_011516940.2 XP_011515242.1 8 - 9 1532 88
XM_017013241.1 XP_016868730.1 17 - 18 2879 88
XM_017013242.1 XP_016868731.1 18 - 19 3240 88
XM_017013243.1 XP_016868732.1 9 - 10 1992 88
GenBank mRNA AB020677.2 - 12 - 13 2173 88
BC133023.1 - 15 - 16 2573 88
BC144226.1 - 15 - 16 2573 88

View Full Product Details