The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252049.1 | NP_001238978.1 | 3 - 4 | 395 | 94 |
NM_001252053.1 | NP_001238982.1 | 3 - 4 | 395 | 94 | |
NM_005389.2 | NP_005380.2 | 3 - 4 | 395 | 94 | |
XM_011535868.2 | XP_011534170.1 | 3 - 4 | 596 | 94 | |
GenBank mRNA | AK129778.1 | - | 3 - 4 | 311 | 94 |
AK289724.1 | - | 3 - 4 | 363 | 94 | |
BC007501.2 | - | 3 - 4 | 337 | 94 | |
BC008748.2 | - | 3 - 4 | 273 | 94 | |
CX786396.1 | - | 3 - 4 | 281 | 94 | |
D13892.1 | - | 3 - 4 | 338 | 94 | |
D25545.1 | - | 3 - 4 | 261 | 94 | |
D25546.1 | - | 3 - 4 | 261 | 94 | |
D25547.1 | - | 3 - 4 | 261 | 94 | |
DB478200.1 | - | 3 - 4 | 385 | 94 | |
DC339361.1 | - | 3 - 4 | 391 | 94 | |
DC421281.1 | - | 3 - 4 | 385 | 94 | |
M93008.1 | - | 3 - 4 | 225 | 94 |