The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320144.1 | NP_001307073.1 | 3 - 4 | 726 | 76 |
GenBank mRNA | AL049667.1 | - | 3 - 4 | 579 | 76 |
AL558384.3 | - | 3 - 4 | 726 | 76 |