The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308020.1 | NP_001294949.1 | 11 - 12 | 1317 | 89 |
NM_002647.3 | NP_002638.2 | 12 - 13 | 1506 | 89 | |
XM_011526031.2 | XP_011524333.1 | 13 - 14 | 1671 | 89 | |
GenBank mRNA | AK298517.1 | - | 11 - 12 | 1261 | 89 |
BC033004.2 | - | 12 - 13 | 1456 | 89 | |
BC053651.1 | - | 12 - 13 | 1436 | 89 | |
Z46973.1 | - | 12 - 13 | 1459 | 89 |