The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308020.1 | NP_001294949.1 | 17 - 18 | 1946 | 112 |
NM_002647.3 | NP_002638.2 | 18 - 19 | 2135 | 112 | |
XM_011526031.2 | XP_011524333.1 | 19 - 20 | 2300 | 112 | |
GenBank mRNA | AK022653.1 | - | 2 - 3 | 144 | 112 |
AK090567.1 | - | 5 - 6 | 2669 | 112 | |
AK298517.1 | - | 17 - 18 | 1890 | 112 | |
BC033004.2 | - | 18 - 19 | 2085 | 112 | |
BC053651.1 | - | 18 - 19 | 2065 | 112 | |
Z46973.1 | - | 18 - 19 | 2088 | 112 |