Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGSF11
Entrez Gene ID
152404
Gene Name
immunoglobulin superfamily member 11
Gene Aliases
BT-IgSF, CT119, CXADRL1, Igsf13, VSIG3
Chromosome Location
Chr.3: 118900630 - 119146068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015887.1 NP_001015887.1 1 - 2 297 61
GenBank mRNA AB071618.1 - 1 - 2 287 61
AB079879.1 - 1 - 2 50 61
AK122611.1 - 1 - 2 260 61
AK296503.1 - 1 - 2 196 61
AY358141.1 - 1 - 2 338 61

View Full Product Details