Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HES2
Entrez Gene ID
54626
Gene Name
hes family bHLH transcription factor 2
Gene Aliases
bHLHb40
Chromosome Location
Chr.1: 6415232 - 6419919 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC012091.1 - 1 - 2 197 57

View Full Product Details