Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYND11
Entrez Gene ID
10771
Gene Name
zinc finger MYND-type containing 11
Gene Aliases
BRAM1, BS69, MRD30
Chromosome Location
Chr.10: 134340 - 254637 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202464.1 NP_001189393.1 11 - 12 1165 79
NM_001202465.1 NP_001189394.1 10 - 11 1405 79
NM_001202466.1 NP_001189395.1 11 - 12 1495 79
NM_001202467.1 NP_001189396.1 10 - 11 1089 79
NM_001202468.1 NP_001189397.1 11 - 12 1251 79
NM_006624.5 NP_006615.2 12 - 13 1573 79
NM_212479.3 NP_997644.2 12 - 13 1657 79
XM_005252359.4 XP_005252416.1 12 - 13 1413 79
XM_005252361.3 XP_005252418.1 11 - 12 1334 79
XM_005252362.2 XP_005252419.1 11 - 12 1498 79
XM_006717376.2 XP_006717439.1 11 - 12 1536 79
XM_017015587.1 XP_016871076.1 12 - 13 1326 79
XM_017015588.1 XP_016871077.1 12 - 13 1562 79
XM_017015589.1 XP_016871078.1 12 - 13 1403 79
XM_017015590.1 XP_016871079.1 12 - 13 1860 79
XM_017015591.1 XP_016871080.1 13 - 14 1373 79
XM_017015592.1 XP_016871081.1 11 - 12 1241 79
XM_017015593.1 XP_016871082.1 11 - 12 1681 79
XM_017015594.1 XP_016871083.1 11 - 12 1169 79
GenBank mRNA AK294469.1 - 10 - 11 1087 79
AK294781.1 - 11 - 12 1177 79
AK296427.1 - 10 - 11 1061 79
AK296518.1 - 7 - 8 748 79
AK312570.1 - 12 - 13 1310 79
AY732489.1 - 8 - 9 872 79
BC034784.1 - 12 - 13 1438 79
BC091489.1 - 11 - 12 1165 79
DQ335452.1 - 11 - 12 1232 79
DQ335453.1 - 10 - 11 1070 79
DQ335454.1 - 10 - 11 1070 79
DQ335455.1 - 11 - 12 1232 79
X86098.1 - 12 - 13 1356 79

View Full Product Details