Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYND11
Entrez Gene ID
10771
Gene Name
zinc finger MYND-type containing 11
Gene Aliases
BRAM1, BS69, MRD30
Chromosome Location
Chr.10: 134340 - 254637 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202464.1 NP_001189393.1 5 - 6 539 86
NM_001202466.1 NP_001189395.1 5 - 6 872 86
NM_001202467.1 NP_001189396.1 4 - 5 463 86
NM_001202468.1 NP_001189397.1 5 - 6 625 86
NM_006624.5 NP_006615.2 6 - 7 947 86
NM_212479.3 NP_997644.2 6 - 7 1034 86
XM_005252359.4 XP_005252416.1 6 - 7 787 86
XM_005252361.3 XP_005252418.1 5 - 6 708 86
XM_005252362.2 XP_005252419.1 5 - 6 872 86
XM_006717376.2 XP_006717439.1 5 - 6 910 86
XM_017015587.1 XP_016871076.1 6 - 7 700 86
XM_017015588.1 XP_016871077.1 6 - 7 936 86
XM_017015589.1 XP_016871078.1 6 - 7 777 86
XM_017015590.1 XP_016871079.1 6 - 7 1234 86
XM_017015591.1 XP_016871080.1 7 - 8 747 86
XM_017015592.1 XP_016871081.1 5 - 6 615 86
XM_017015593.1 XP_016871082.1 5 - 6 1055 86
XM_017015594.1 XP_016871083.1 5 - 6 543 86
GenBank mRNA AK294781.1 - 5 - 6 554 86
AK296427.1 - 4 - 5 435 86
AK312570.1 - 6 - 7 684 86
AY732489.1 - 3 - 4 324 86
BC012586.2 - 5 - 6 626 86
BC034784.1 - 6 - 7 815 86
BC091489.1 - 5 - 6 539 86
DQ335452.1 - 5 - 6 606 86
DQ335453.1 - 4 - 5 444 86
DQ335454.1 - 4 - 5 444 86
DQ335455.1 - 5 - 6 606 86
X86098.1 - 6 - 7 730 86

View Full Product Details