Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEST3
Entrez Gene ID
144453
Gene Name
bestrophin 3
Gene Aliases
VMD2L3
Chromosome Location
Chr.12: 69643508 - 69699416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282614.1 NP_001269543.1 1 - 2 204 114
NM_032735.2 NP_116124.2 1 - 2 204 114
GenBank mRNA AF440758.1 - 1 - 2 149 114
AK096459.1 - 1 - 2 204 115
DA767581.1 - 1 - 2 204 115

View Full Product Details