Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBL7
Entrez Gene ID
84993
Gene Name
ubiquitin like 7
Gene Aliases
BMSC-UbP, TCBA1
Chromosome Location
Chr.15: 74445977 - 74461188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286739.1 NP_001273668.1 6 - 7 665 70
NM_001286740.1 NP_001273669.1 5 - 6 530 70
NM_001286741.1 NP_001273670.1 5 - 6 1020 70
NM_001286742.1 NP_001273671.1 6 - 7 762 70
NM_032907.4 NP_116296.1 5 - 6 651 70
NM_201265.1 NP_957717.1 5 - 6 580 70
XM_011522124.2 XP_011520426.1 6 - 7 867 70
GenBank mRNA AY037166.1 - 5 - 6 554 70
BC007913.2 - 5 - 6 934 70
BC030055.1 - 6 - 7 628 70
BC033919.1 - 6 - 7 669 70
BI551787.1 - 5 - 6 637 70
BI552240.1 - 5 - 6 586 70

View Full Product Details