The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286739.1 | NP_001273668.1 | 8 - 9 | 858 | 64 |
NM_001286740.1 | NP_001273669.1 | 7 - 8 | 723 | 64 | |
NM_001286741.1 | NP_001273670.1 | 7 - 8 | 1213 | 64 | |
NM_001286742.1 | NP_001273671.1 | 8 - 9 | 955 | 64 | |
NM_032907.4 | NP_116296.1 | 7 - 8 | 844 | 64 | |
NM_201265.1 | NP_957717.1 | 7 - 8 | 773 | 64 | |
XM_011522124.2 | XP_011520426.1 | 8 - 9 | 1060 | 64 | |
GenBank mRNA | AY037166.1 | - | 7 - 8 | 747 | 64 |
BC007913.2 | - | 7 - 8 | 1127 | 64 | |
BC030055.1 | - | 8 - 9 | 821 | 64 | |
BC033919.1 | - | 8 - 9 | 862 | 64 |