The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024334.2 | NP_077310.1 | 10 - 11 | 1132 | 114 |
XM_017007176.1 | XP_016862665.1 | 11 - 12 | 1254 | 114 | |
GenBank mRNA | AK027466.1 | - | 4 - 5 | 362 | 114 |
AK027757.1 | - | 10 - 11 | 1058 | 114 | |
AK027827.1 | - | 5 - 6 | 415 | 114 | |
AK027877.1 | - | 10 - 11 | 996 | 114 | |
AK075010.1 | - | 10 - 11 | 983 | 114 | |
AK225589.1 | - | 10 - 11 | 1055 | 114 | |
AL136916.1 | - | 10 - 11 | 1026 | 114 | |
AY358625.1 | - | 10 - 11 | 895 | 114 | |
BC003125.1 | - | 10 - 11 | 1027 | 114 | |
BC008054.2 | - | 8 - 9 | 622 | 114 | |
BC011719.2 | - | 10 - 11 | 1027 | 114 | |
KU178769.1 | - | 9 - 10 | 878 | 114 | |
KU178770.1 | - | 8 - 9 | 1452 | 114 |