Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP35
Entrez Gene ID
57558
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
-
Chromosome Location
Chr.11: 78188812 - 78233651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020798.2 NP_065849.1 4 - 5 1289 106
XM_011545187.2 XP_011543489.1 4 - 5 432 106
XM_011545188.2 XP_011543490.1 2 - 3 215 106
XM_017018052.1 XP_016873541.1 4 - 5 1286 106
XM_017018053.1 XP_016873542.1 3 - 4 281 106
GenBank mRNA AB037793.1 - 1 - 2 752 106
AJ586137.1 - 3 - 4 930 106
AK307875.1 - 3 - 4 515 106

View Full Product Details