Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP35
Entrez Gene ID
57558
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
-
Chromosome Location
Chr.11: 78188812 - 78233651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020798.2 NP_065849.1 6 - 7 1558 69
XM_011545187.2 XP_011543489.1 6 - 7 701 69
XM_011545188.2 XP_011543490.1 4 - 5 484 69
XM_017018052.1 XP_016873541.1 6 - 7 1555 69
XM_017018053.1 XP_016873542.1 5 - 6 550 69
GenBank mRNA AB037793.1 - 3 - 4 1021 69
AJ586137.1 - 5 - 6 1199 69
AK307875.1 - 5 - 6 784 69
BC131489.1 - 4 - 5 521 69

View Full Product Details