The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000812.3 | NP_000803.2 | 2 - 3 | 548 | 84 |
XM_017007986.1 | XP_016863475.1 | 2 - 3 | 273 | 84 | |
GenBank mRNA | AK225167.1 | - | 2 - 3 | 225 | 84 |
AK225686.1 | - | 2 - 3 | 252 | 84 | |
AK296023.1 | - | 2 - 3 | 441 | 84 | |
AK307120.1 | - | 2 - 3 | 275 | 84 | |
AK312720.1 | - | 2 - 3 | 194 | 84 | |
BC022449.1 | - | 2 - 3 | 252 | 84 | |
KJ534841.1 | - | 2 - 3 | 174 | 84 | |
X14767.1 | - | 2 - 3 | 205 | 84 |