Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018313.4 NP_060783.3 10 - 11 1107 74
XM_005265279.4 XP_005265336.1 11 - 12 1335 74
XM_005265280.3 XP_005265337.1 10 - 11 1152 74
XM_005265282.3 XP_005265339.1 11 - 12 1294 74
XM_005265283.3 XP_005265340.1 10 - 11 1149 74
XM_011533900.2 XP_011532202.1 13 - 14 1501 74
XM_011533902.2 XP_011532204.1 12 - 13 1433 74
XM_011533903.2 XP_011532205.1 12 - 13 1438 74
XM_017006725.1 XP_016862214.1 12 - 13 1408 74
XM_017006726.1 XP_016862215.1 13 - 14 1518 74
XM_017006727.1 XP_016862216.1 13 - 14 1463 74
XM_017006728.1 XP_016862217.1 12 - 13 1459 74
XM_017006729.1 XP_016862218.1 12 - 13 1867 74
XM_017006730.1 XP_016862219.1 13 - 14 1454 74
XM_017006731.1 XP_016862220.1 12 - 13 1437 74
XM_017006732.1 XP_016862221.1 12 - 13 1433 74
XM_017006733.1 XP_016862222.1 12 - 13 1827 74
XM_017006734.1 XP_016862223.1 12 - 13 1400 74
XM_017006735.1 XP_016862224.1 12 - 13 1866 74
XM_017006736.1 XP_016862225.1 12 - 13 1867 74
XM_017006737.1 XP_016862226.1 11 - 12 1741 74
XM_017006738.1 XP_016862227.1 11 - 12 1362 74
XM_017006739.1 XP_016862228.1 11 - 12 1738 74
XM_017006740.1 XP_016862229.1 11 - 12 1279 74
XM_017006741.1 XP_016862230.1 12 - 13 1415 74
XM_017006742.1 XP_016862231.1 12 - 13 1438 74
XM_017006743.1 XP_016862232.1 12 - 13 1867 74
XM_017006744.1 XP_016862233.1 12 - 13 1404 74
XM_017006745.1 XP_016862234.1 11 - 12 1713 74
XM_017006746.1 XP_016862235.1 11 - 12 1710 74
XM_017006748.1 XP_016862237.1 11 - 12 1713 74
XM_017006749.1 XP_016862238.1 11 - 12 1279 74
XM_017006750.1 XP_016862239.1 10 - 11 1167 74
XM_017006751.1 XP_016862240.1 12 - 13 1415 74
XM_017006752.1 XP_016862241.1 12 - 13 1867 74
XM_017006753.1 XP_016862242.1 12 - 13 1866 74
XM_017006754.1 XP_016862243.1 12 - 13 1433 74
XM_017006755.1 XP_016862244.1 12 - 13 1415 74
XM_017006756.1 XP_016862245.1 12 - 13 1415 74
XM_017006757.1 XP_016862246.1 10 - 11 1138 74
XM_017006758.1 XP_016862247.1 10 - 11 1269 74
XM_017006759.1 XP_016862248.1 12 - 13 1415 74
XM_017006760.1 XP_016862249.1 12 - 13 1433 74
XM_017006761.1 XP_016862250.1 12 - 13 1442 74
XM_017006762.1 XP_016862251.1 12 - 13 1433 74
XM_017006763.1 XP_016862252.1 13 - 14 1492 74
XM_017006764.1 XP_016862253.1 12 - 13 1415 74
XM_017006765.1 XP_016862254.1 11 - 12 1294 74
XM_017006766.1 XP_016862255.1 12 - 13 1422 74
GenBank mRNA AF177387.1 - 9 - 10 996 74
AF197569.1 - 10 - 11 1089 74
AF225871.1 - 9 - 10 996 74
AF225872.1 - 9 - 10 996 74
AK056541.1 - 8 - 9 825 74
AY281068.1 - 9 - 10 995 74
BC115009.1 - 9 - 10 1011 74
BC115010.1 - 9 - 10 1012 74
BC115011.1 - 9 - 10 1012 74
BC129934.1 - 9 - 10 996 74
BC129935.1 - 9 - 10 996 74
BU171600.1 - 2 - 3 202 74

View Full Product Details