Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018313.4 NP_060783.3 13 - 14 1557 89
XM_005265279.4 XP_005265336.1 14 - 15 1785 89
XM_005265280.3 XP_005265337.1 13 - 14 1602 89
XM_005265282.3 XP_005265339.1 14 - 15 1744 89
XM_005265283.3 XP_005265340.1 13 - 14 1599 89
XM_011533900.2 XP_011532202.1 16 - 17 1951 89
XM_011533902.2 XP_011532204.1 15 - 16 1883 89
XM_011533903.2 XP_011532205.1 15 - 16 1888 89
XM_017006725.1 XP_016862214.1 15 - 16 1858 89
XM_017006726.1 XP_016862215.1 16 - 17 1968 89
XM_017006727.1 XP_016862216.1 16 - 17 1913 89
XM_017006728.1 XP_016862217.1 15 - 16 1909 89
XM_017006729.1 XP_016862218.1 15 - 16 2317 89
XM_017006730.1 XP_016862219.1 16 - 17 1904 89
XM_017006731.1 XP_016862220.1 15 - 16 1887 89
XM_017006732.1 XP_016862221.1 15 - 16 1883 89
XM_017006733.1 XP_016862222.1 15 - 16 2277 89
XM_017006734.1 XP_016862223.1 15 - 16 1850 89
XM_017006735.1 XP_016862224.1 15 - 16 2316 89
XM_017006736.1 XP_016862225.1 15 - 16 2317 89
XM_017006737.1 XP_016862226.1 14 - 15 2191 89
XM_017006738.1 XP_016862227.1 14 - 15 1812 89
XM_017006739.1 XP_016862228.1 14 - 15 2188 89
XM_017006740.1 XP_016862229.1 14 - 15 1729 89
XM_017006741.1 XP_016862230.1 15 - 16 1865 89
XM_017006742.1 XP_016862231.1 15 - 16 1888 89
XM_017006743.1 XP_016862232.1 15 - 16 2317 89
XM_017006744.1 XP_016862233.1 15 - 16 1854 89
XM_017006745.1 XP_016862234.1 14 - 15 2163 89
XM_017006746.1 XP_016862235.1 14 - 15 2160 89
XM_017006747.1 XP_016862236.1 14 - 15 2222 89
XM_017006748.1 XP_016862237.1 14 - 15 2163 89
XM_017006749.1 XP_016862238.1 14 - 15 1729 89
XM_017006750.1 XP_016862239.1 13 - 14 1617 89
XM_017006751.1 XP_016862240.1 15 - 16 1865 89
XM_017006752.1 XP_016862241.1 15 - 16 2317 89
XM_017006753.1 XP_016862242.1 15 - 16 2316 89
XM_017006754.1 XP_016862243.1 15 - 16 1883 89
XM_017006755.1 XP_016862244.1 15 - 16 1865 89
XM_017006756.1 XP_016862245.1 15 - 16 1865 89
XM_017006757.1 XP_016862246.1 13 - 14 1588 89
XM_017006758.1 XP_016862247.1 13 - 14 1719 89
XM_017006759.1 XP_016862248.1 15 - 16 1865 89
XM_017006760.1 XP_016862249.1 15 - 16 1883 89
XM_017006761.1 XP_016862250.1 15 - 16 1892 89
XM_017006762.1 XP_016862251.1 15 - 16 1883 89
XM_017006763.1 XP_016862252.1 16 - 17 1942 89
XM_017006764.1 XP_016862253.1 15 - 16 1865 89
XM_017006765.1 XP_016862254.1 14 - 15 1744 89
XM_017006766.1 XP_016862255.1 15 - 16 1872 89
GenBank mRNA AF177387.1 - 12 - 13 1446 89
AF197569.1 - 13 - 14 1539 89
AF225870.1 - 11 - 12 1350 89
AF225871.1 - 12 - 13 1446 89
AF225872.1 - 12 - 13 1446 89
AK056541.1 - 11 - 12 1275 89
AY281068.1 - 12 - 13 1445 89
BC115009.1 - 12 - 13 1461 89
BC115010.1 - 12 - 13 1462 89
BC115011.1 - 12 - 13 1462 89
BC129934.1 - 12 - 13 1446 89
BC129935.1 - 12 - 13 1446 89
BU171600.1 - 5 - 6 652 89

View Full Product Details