Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018313.4 NP_060783.3 28 - 29 4476 115
XM_005265279.4 XP_005265336.1 31 - 32 5070 115
XM_005265280.3 XP_005265337.1 30 - 31 4887 115
XM_005265282.3 XP_005265339.1 31 - 32 5029 115
XM_005265283.3 XP_005265340.1 30 - 31 4884 115
XM_011533900.2 XP_011532202.1 33 - 34 5236 115
XM_011533902.2 XP_011532204.1 32 - 33 5168 115
XM_011533903.2 XP_011532205.1 32 - 33 5173 115
XM_017006725.1 XP_016862214.1 32 - 33 5143 115
XM_017006726.1 XP_016862215.1 33 - 34 5253 115
XM_017006727.1 XP_016862216.1 33 - 34 5198 115
XM_017006728.1 XP_016862217.1 32 - 33 5194 115
XM_017006729.1 XP_016862218.1 32 - 33 5599 115
XM_017006730.1 XP_016862219.1 33 - 34 5189 115
XM_017006731.1 XP_016862220.1 32 - 33 5172 115
XM_017006732.1 XP_016862221.1 32 - 33 5165 115
XM_017006733.1 XP_016862222.1 32 - 33 5562 115
XM_017006734.1 XP_016862223.1 32 - 33 5135 115
XM_017006735.1 XP_016862224.1 31 - 32 5556 115
XM_017006736.1 XP_016862225.1 31 - 32 5554 115
XM_017006737.1 XP_016862226.1 31 - 32 5476 115
XM_017006738.1 XP_016862227.1 31 - 32 5097 115
XM_017006739.1 XP_016862228.1 31 - 32 5473 115
XM_017006740.1 XP_016862229.1 31 - 32 5014 115
XM_017006741.1 XP_016862230.1 31 - 32 5105 115
XM_017006742.1 XP_016862231.1 31 - 32 5128 115
XM_017006743.1 XP_016862232.1 32 - 33 5527 115
XM_017006744.1 XP_016862233.1 31 - 32 5094 115
XM_017006745.1 XP_016862234.1 31 - 32 5448 115
XM_017006746.1 XP_016862235.1 31 - 32 5445 115
XM_017006747.1 XP_016862236.1 31 - 32 5507 115
XM_017006748.1 XP_016862237.1 30 - 31 5403 115
XM_017006749.1 XP_016862238.1 30 - 31 4969 115
XM_017006750.1 XP_016862239.1 29 - 30 4857 115
XM_017006751.1 XP_016862240.1 31 - 32 5030 115
XM_017006752.1 XP_016862241.1 31 - 32 5446 115
XM_017006753.1 XP_016862242.1 31 - 32 5436 115
XM_017006754.1 XP_016862243.1 31 - 32 5012 115
XM_017006755.1 XP_016862244.1 30 - 31 4940 115
XM_017006756.1 XP_016862245.1 30 - 31 4937 115
XM_017006757.1 XP_016862246.1 28 - 29 4663 115
XM_017006758.1 XP_016862247.1 28 - 29 4794 115
XM_017006759.1 XP_016862248.1 30 - 31 4865 115
XM_017006760.1 XP_016862249.1 30 - 31 4847 115
XM_017006761.1 XP_016862250.1 30 - 31 4856 115
XM_017006762.1 XP_016862251.1 30 - 31 4844 115
XM_017006763.1 XP_016862252.1 31 - 32 4906 115
XM_017006764.1 XP_016862253.1 29 - 30 4784 115
XM_017006765.1 XP_016862254.1 28 - 29 4663 115
XM_017006766.1 XP_016862255.1 29 - 30 4716 115
GenBank mRNA AF177387.1 - 27 - 28 4521 115
AF197569.1 - 28 - 29 4458 115
AF225870.1 - 26 - 27 4425 115
AF225871.1 - 26 - 27 4306 115
AF225872.1 - 26 - 27 4365 115
AK001507.1 - 4 - 5 548 115
AY281068.1 - 28 - 29 4685 115
BC015323.1 - 5 - 6 812 115
BC089409.1 - 6 - 7 893 115
BC115010.1 - 27 - 28 4462 115
BC115011.1 - 27 - 28 4537 115
BC129934.1 - 28 - 29 4575 115
BC129935.1 - 27 - 28 4410 115

View Full Product Details