Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018313.4 NP_060783.3 2 - 3 252 92
XM_005265279.4 XP_005265336.1 3 - 4 480 92
XM_005265280.3 XP_005265337.1 2 - 3 297 92
XM_005265282.3 XP_005265339.1 3 - 4 439 92
XM_005265283.3 XP_005265340.1 2 - 3 294 92
XM_011533900.2 XP_011532202.1 5 - 6 646 92
XM_011533902.2 XP_011532204.1 4 - 5 578 92
XM_011533903.2 XP_011532205.1 4 - 5 583 92
XM_017006725.1 XP_016862214.1 4 - 5 526 92
XM_017006726.1 XP_016862215.1 5 - 6 636 92
XM_017006727.1 XP_016862216.1 5 - 6 581 92
XM_017006728.1 XP_016862217.1 4 - 5 577 92
XM_017006729.1 XP_016862218.1 4 - 5 985 92
XM_017006730.1 XP_016862219.1 5 - 6 572 92
XM_017006731.1 XP_016862220.1 4 - 5 555 92
XM_017006732.1 XP_016862221.1 4 - 5 578 92
XM_017006733.1 XP_016862222.1 4 - 5 972 92
XM_017006734.1 XP_016862223.1 4 - 5 545 92
XM_017006735.1 XP_016862224.1 4 - 5 984 92
XM_017006736.1 XP_016862225.1 4 - 5 985 92
XM_017006737.1 XP_016862226.1 3 - 4 859 92
XM_017006738.1 XP_016862227.1 3 - 4 480 92
XM_017006739.1 XP_016862228.1 3 - 4 856 92
XM_017006740.1 XP_016862229.1 3 - 4 397 92
XM_017006741.1 XP_016862230.1 4 - 5 560 92
XM_017006742.1 XP_016862231.1 4 - 5 583 92
XM_017006743.1 XP_016862232.1 4 - 5 985 92
XM_017006744.1 XP_016862233.1 4 - 5 549 92
XM_017006745.1 XP_016862234.1 3 - 4 858 92
XM_017006746.1 XP_016862235.1 3 - 4 855 92
XM_017006747.1 XP_016862236.1 4 - 5 986 92
XM_017006748.1 XP_016862237.1 3 - 4 858 92
XM_017006749.1 XP_016862238.1 3 - 4 424 92
XM_017006750.1 XP_016862239.1 2 - 3 312 92
XM_017006751.1 XP_016862240.1 4 - 5 560 92
XM_017006752.1 XP_016862241.1 4 - 5 985 92
XM_017006753.1 XP_016862242.1 4 - 5 984 92
XM_017006754.1 XP_016862243.1 4 - 5 578 92
XM_017006755.1 XP_016862244.1 4 - 5 560 92
XM_017006756.1 XP_016862245.1 4 - 5 560 92
XM_017006757.1 XP_016862246.1 2 - 3 283 92
XM_017006758.1 XP_016862247.1 2 - 3 414 92
XM_017006759.1 XP_016862248.1 4 - 5 560 92
XM_017006760.1 XP_016862249.1 4 - 5 578 92
XM_017006761.1 XP_016862250.1 4 - 5 587 92
XM_017006762.1 XP_016862251.1 4 - 5 578 92
XM_017006763.1 XP_016862252.1 5 - 6 637 92
XM_017006764.1 XP_016862253.1 4 - 5 560 92
XM_017006765.1 XP_016862254.1 3 - 4 439 92
XM_017006766.1 XP_016862255.1 4 - 5 567 92
GenBank mRNA AF177387.1 - 1 - 2 141 92
AF197569.1 - 2 - 3 234 92
AF225870.1 - 1 - 2 141 92
AF225871.1 - 1 - 2 141 92
AF225872.1 - 1 - 2 141 92
AY281068.1 - 1 - 2 140 92
BC115009.1 - 1 - 2 156 92
BC115011.1 - 1 - 2 157 92
BC129934.1 - 1 - 2 141 92
BC129935.1 - 1 - 2 141 92
BE672944.1 - 2 - 3 252 92

View Full Product Details