Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018313.4 NP_060783.3 7 - 8 824 122
XM_005265279.4 XP_005265336.1 8 - 9 1052 122
XM_005265280.3 XP_005265337.1 7 - 8 869 122
XM_005265282.3 XP_005265339.1 8 - 9 1011 122
XM_005265283.3 XP_005265340.1 7 - 8 866 122
XM_011533900.2 XP_011532202.1 10 - 11 1218 122
XM_011533902.2 XP_011532204.1 9 - 10 1150 122
XM_011533903.2 XP_011532205.1 9 - 10 1155 122
XM_017006725.1 XP_016862214.1 9 - 10 1125 122
XM_017006726.1 XP_016862215.1 10 - 11 1235 122
XM_017006727.1 XP_016862216.1 10 - 11 1180 122
XM_017006728.1 XP_016862217.1 9 - 10 1176 122
XM_017006729.1 XP_016862218.1 9 - 10 1584 122
XM_017006730.1 XP_016862219.1 10 - 11 1171 122
XM_017006731.1 XP_016862220.1 9 - 10 1154 122
XM_017006732.1 XP_016862221.1 9 - 10 1150 122
XM_017006733.1 XP_016862222.1 9 - 10 1544 122
XM_017006734.1 XP_016862223.1 9 - 10 1117 122
XM_017006735.1 XP_016862224.1 9 - 10 1583 122
XM_017006736.1 XP_016862225.1 9 - 10 1584 122
XM_017006737.1 XP_016862226.1 8 - 9 1458 122
XM_017006738.1 XP_016862227.1 8 - 9 1079 122
XM_017006739.1 XP_016862228.1 8 - 9 1455 122
XM_017006740.1 XP_016862229.1 8 - 9 996 122
XM_017006741.1 XP_016862230.1 9 - 10 1132 122
XM_017006742.1 XP_016862231.1 9 - 10 1155 122
XM_017006743.1 XP_016862232.1 9 - 10 1584 122
XM_017006744.1 XP_016862233.1 9 - 10 1121 122
XM_017006745.1 XP_016862234.1 8 - 9 1430 122
XM_017006746.1 XP_016862235.1 8 - 9 1427 122
XM_017006747.1 XP_016862236.1 9 - 10 1585 122
XM_017006748.1 XP_016862237.1 8 - 9 1430 122
XM_017006749.1 XP_016862238.1 8 - 9 996 122
XM_017006750.1 XP_016862239.1 7 - 8 884 122
XM_017006751.1 XP_016862240.1 9 - 10 1132 122
XM_017006752.1 XP_016862241.1 9 - 10 1584 122
XM_017006753.1 XP_016862242.1 9 - 10 1583 122
XM_017006754.1 XP_016862243.1 9 - 10 1150 122
XM_017006755.1 XP_016862244.1 9 - 10 1132 122
XM_017006756.1 XP_016862245.1 9 - 10 1132 122
XM_017006757.1 XP_016862246.1 7 - 8 855 122
XM_017006758.1 XP_016862247.1 7 - 8 986 122
XM_017006759.1 XP_016862248.1 9 - 10 1132 122
XM_017006760.1 XP_016862249.1 9 - 10 1150 122
XM_017006761.1 XP_016862250.1 9 - 10 1159 122
XM_017006762.1 XP_016862251.1 9 - 10 1150 122
XM_017006763.1 XP_016862252.1 10 - 11 1209 122
XM_017006764.1 XP_016862253.1 9 - 10 1132 122
XM_017006765.1 XP_016862254.1 8 - 9 1011 122
XM_017006766.1 XP_016862255.1 9 - 10 1139 122
GenBank mRNA AF177387.1 - 6 - 7 713 122
AF197569.1 - 7 - 8 806 122
AF225870.1 - 6 - 7 713 122
AF225871.1 - 6 - 7 713 122
AF225872.1 - 6 - 7 713 122
AK056541.1 - 5 - 6 542 122
AY281068.1 - 6 - 7 712 122
BC115009.1 - 6 - 7 728 122
BC115010.1 - 6 - 7 729 122
BC129934.1 - 6 - 7 713 122
BC129935.1 - 6 - 7 713 122

View Full Product Details