Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALKBH6
Entrez Gene ID
84964
Gene Name
alkB homolog 6
Gene Aliases
ABH6
Chromosome Location
Chr.19: 36009116 - 36014243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297701.1 NP_001284630.1 2 - 3 143 65
XM_005259357.4 XP_005259414.1 1 - 2 111 65
GenBank mRNA AB273714.1 - 2 - 3 143 65
BM713594.1 - 2 - 3 139 65

View Full Product Details