The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195248.1 | NP_001182177.1 | 7 - 8 | 960 | 131 |
NM_001195249.1 | NP_001182178.1 | 7 - 8 | 1129 | 131 | |
NM_001195250.1 | NP_001182179.1 | 6 - 7 | 798 | 131 | |
NM_001195252.1 | NP_001182181.1 | 7 - 8 | 744 | 131 | |
NM_001195254.1 | NP_001182183.1 | 6 - 7 | 820 | 131 | |
NM_175073.2 | NP_778243.1 | 8 - 9 | 1066 | 131 | |
NR_036576.1 | 7 - 8 | 955 | 131 | ||
NR_036577.1 | 7 - 8 | 845 | 131 | ||
NR_036578.1 | 7 - 8 | 977 | 131 | ||
NR_036579.1 | 7 - 8 | 1124 | 131 | ||
XM_006716791.3 | XP_006716854.1 | 7 - 8 | 968 | 131 | |
XM_006716792.2 | XP_006716855.1 | 7 - 8 | 986 | 131 | |
XM_011517938.1 | XP_011516240.1 | 6 - 7 | 944 | 131 | |
XM_011517939.2 | XP_011516241.1 | 6 - 7 | 899 | 131 | |
XM_017014837.1 | XP_016870326.1 | 6 - 7 | 1049 | 131 | |
GenBank mRNA | AK000164.1 | - | 6 - 7 | 963 | 131 |
AK131046.1 | - | 7 - 8 | 912 | 131 | |
AY040777.1 | - | 7 - 8 | 935 | 131 | |
AY208829.1 | - | 7 - 8 | 935 | 131 | |
AY208830.1 | - | 6 - 7 | 798 | 131 | |
AY208831.1 | - | 7 - 8 | 710 | 131 | |
AY208832.1 | - | 6 - 7 | 773 | 131 | |
AY208833.1 | - | 6 - 7 | 773 | 131 | |
AY208834.1 | - | 6 - 7 | 874 | 131 | |
AY208835.1 | - | 7 - 8 | 915 | 131 | |
AY208836.1 | - | 7 - 8 | 1075 | 131 | |
AY208837.1 | - | 8 - 9 | 1032 | 131 | |
AY208838.1 | - | 6 - 7 | 891 | 131 | |
AY208839.1 | - | 7 - 8 | 1069 | 131 | |
AY208840.1 | - | 7 - 8 | 932 | 131 | |
AY208841.1 | - | 6 - 7 | 769 | 131 | |
AY302067.1 | - | 6 - 7 | 1078 | 131 | |
AY302068.1 | - | 6 - 7 | 1073 | 131 | |
AY302069.1 | - | 6 - 7 | 963 | 131 | |
AY302070.1 | - | 6 - 7 | 958 | 131 | |
AY302071.1 | - | 5 - 6 | 851 | 131 | |
AY302072.1 | - | 5 - 6 | 846 | 131 | |
AY302073.1 | - | 5 - 6 | 736 | 131 | |
AY302074.1 | - | 5 - 6 | 731 | 131 | |
BC001628.1 | - | 6 - 7 | 783 | 131 | |
BC032650.1 | - | 7 - 8 | 1001 | 131 | |
BC068107.1 | - | 8 - 9 | 1041 | 131 | |
BC104881.1 | - | 7 - 8 | 974 | 131 |