The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024605.3 | NP_078881.3 | 4 - 5 | 379 | 163 |
XM_005263215.3 | XP_005263272.1 | 4 - 5 | 615 | 163 | |
GenBank mRNA | AB050785.1 | - | 4 - 5 | 379 | 163 |
BC109029.2 | - | 4 - 5 | 369 | 163 | |
BC109030.1 | - | 4 - 5 | 368 | 163 | |
BC126899.1 | - | 4 - 5 | 375 | 163 | |
BC128055.1 | - | 4 - 5 | 379 | 163 |