Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYN3
Entrez Gene ID
8224
Gene Name
synapsin III
Gene Aliases
-
Chromosome Location
Chr.22: 32508022 - 33058391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135774.1 NP_001129246.1 8 - 9 1176 74
NM_003490.3 NP_003481.3 7 - 8 1076 74
NM_133633.2 NP_598344.2 7 - 8 1076 74
XM_011530405.2 XP_011528707.1 8 - 9 1712 74
XM_011530406.2 XP_011528708.1 8 - 9 1178 74
XM_011530407.2 XP_011528709.1 8 - 9 1585 74
XM_011530408.1 XP_011528710.1 8 - 9 1176 74
XM_011530410.2 XP_011528712.1 6 - 7 1380 74
XM_017028960.1 XP_016884449.1 9 - 10 2110 74
XM_017028961.1 XP_016884450.1 8 - 9 1502 74
XM_017028962.1 XP_016884451.1 8 - 9 1437 74
XM_017028963.1 XP_016884452.1 8 - 9 1101 74
XM_017028964.1 XP_016884453.1 8 - 9 1173 74
XM_017028965.1 XP_016884454.1 2 - 3 947 74
GenBank mRNA AB209684.1 - 8 - 9 1128 74
AF046873.1 - 7 - 8 953 74
BC075065.2 - 7 - 8 943 74
BC075066.2 - 7 - 8 943 74
BC117460.1 - 7 - 8 941 74
BC143874.1 - 7 - 8 944 74
CR456589.1 - 7 - 8 1008 74

View Full Product Details