Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPST1
Entrez Gene ID
8460
Gene Name
tyrosylprotein sulfotransferase 1
Gene Aliases
TANGO13A
Chromosome Location
Chr.7: 66205272 - 66360451 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003596.3 NP_003587.1 3 - 4 1391 113
XM_017012725.1 XP_016868214.1 4 - 5 1474 113
XM_017012726.1 XP_016868215.1 3 - 4 1387 113
XM_017012727.1 XP_016868216.1 3 - 4 1387 113
GenBank mRNA AF038009.1 - 2 - 3 1120 113
AK307053.1 - 2 - 3 383 113
AK313098.1 - 3 - 4 1329 113
BC013188.1 - 3 - 4 1366 113
CR542080.1 - 2 - 3 1039 113

View Full Product Details