The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014396.3 | NP_055211.2 | 18 - 19 | 1575 | 102 |
NM_080631.3 | NP_542198.2 | 17 - 18 | 1500 | 102 | |
XM_017011988.1 | XP_016867477.1 | 5 - 6 | 508 | 102 | |
GenBank mRNA | AK074325.1 | - | 14 - 15 | 1288 | 102 |
AK299143.1 | - | 15 - 16 | 1321 | 102 | |
AK314556.1 | - | 18 - 19 | 1575 | 102 | |
BC044851.1 | - | 18 - 19 | 1570 | 102 | |
BX648347.1 | - | 16 - 17 | 1300 | 102 | |
U87309.1 | - | 18 - 19 | 1578 | 102 |