The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014396.3 | NP_055211.2 | 6 - 7 | 403 | 103 |
GenBank mRNA | AK074325.1 | - | 2 - 3 | 116 | 103 |
AK299143.1 | - | 3 - 4 | 149 | 103 | |
AK314556.1 | - | 6 - 7 | 403 | 103 | |
BC044851.1 | - | 6 - 7 | 398 | 103 | |
U87309.1 | - | 6 - 7 | 406 | 103 |