The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152743.3 | NP_689956.2 | 10 - 11 | 1682 | 146 |
XM_005249643.4 | XP_005249700.1 | 10 - 11 | 1644 | 146 | |
XM_017011833.1 | XP_016867322.1 | 10 - 11 | 1445 | 146 | |
XM_017011834.1 | XP_016867323.1 | 10 - 11 | 1636 | 146 | |
XM_017011835.1 | XP_016867324.1 | 9 - 10 | 1529 | 146 | |
XM_017011836.1 | XP_016867325.1 | 10 - 11 | 1638 | 146 | |
GenBank mRNA | AL133088.1 | - | 3 - 4 | 359 | 146 |
BC007209.1 | - | 6 - 7 | 917 | 146 | |
BC015632.2 | - | 10 - 11 | 1436 | 146 | |
BC023561.2 | - | 6 - 7 | 677 | 146 |