The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002693.2 | NP_002684.1 | 1 - 2 | 121 | 58 |
GenBank mRNA | AK291281.1 | - | 1 - 2 | 140 | 58 |
BC042571.1 | - | 1 - 2 | 106 | 58 | |
X98093.1 | - | 1 - 2 | 121 | 58 |