Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIP13
Entrez Gene ID
9319
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
16E1BP
Chromosome Location
Chr.5: 892854 - 919348 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004237.3 NP_004228.1 10 - 11 1165 77
XM_011514163.2 XP_011512465.1 10 - 11 1135 77
GenBank mRNA BC000404.2 - 10 - 11 1130 77
BC019294.1 - 10 - 11 1130 77
CR456744.1 - 10 - 11 1020 77
U96131.1 - 10 - 11 1065 77

View Full Product Details