The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001370.1 | NP_001361.1 | 62 - 63 | 10477 | 150 |
XM_006711956.2 | XP_006712019.1 | 62 - 63 | 10582 | 150 | |
XM_011532649.2 | XP_011530951.1 | 62 - 63 | 10431 | 150 | |
XM_011532650.2 | XP_011530952.1 | 62 - 63 | 10432 | 150 | |
XM_011532652.1 | XP_011530954.1 | 61 - 62 | 10351 | 150 | |
XM_011532653.1 | XP_011530955.1 | 61 - 62 | 10330 | 150 | |
XM_011532654.1 | XP_011530956.1 | 61 - 62 | 10210 | 150 | |
XM_011532655.2 | XP_011530957.1 | 58 - 59 | 9499 | 150 | |
XM_011532657.1 | XP_011530959.1 | 56 - 57 | 9238 | 150 | |
XM_017003521.1 | XP_016859010.1 | 60 - 61 | 10204 | 150 | |
XM_017003522.1 | XP_016859011.1 | 57 - 58 | 10303 | 150 | |
XM_017003523.1 | XP_016859012.1 | 57 - 58 | 9470 | 150 | |
GenBank mRNA | AB051484.2 | - | 26 - 27 | 4412 | 150 |
AB290165.1 | - | 28 - 29 | 4616 | 150 | |
AL512706.1 | - | 4 - 5 | 772 | 150 | |
BC117259.1 | - | 15 - 16 | 2609 | 150 |